Product Details

SNP ID
rs6463686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:7345430 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTATTTTTACCTACATAGTTAC[C/T]ATTTCTGGTGTTCTTCGTTCCTTCC
Phenotype
MIM: 609996
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL28A1 PubMed Links

Gene Details

Gene
COL28A1
Gene Name
collagen type XXVIII alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037763.2 Intron NP_001032852.2
XM_011515358.2 Intron XP_011513660.1
XM_011515359.2 Intron XP_011513661.1
XM_011515360.2 Intron XP_011513662.1
XM_011515362.2 Intron XP_011513664.1
XM_011515363.2 Intron XP_011513665.1
XM_011515364.2 Intron XP_011513666.1
XM_011515365.2 Intron XP_011513667.1
XM_011515366.2 Intron XP_011513668.1
XM_017012131.1 Intron XP_016867620.1
XM_017012132.1 Intron XP_016867621.1
XM_017012133.1 Intron XP_016867622.1

View Full Product Details