Product Details

SNP ID
rs10783817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57257471 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTTATACTGCTTGTGACAGCTA[A/G]GGCAGAAAGCAGATAAAATTCCCAT
Phenotype
MIM: 615521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
R3HDM2 PubMed Links

Gene Details

Gene
R3HDM2
Gene Name
R3H domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005268725.3 Intron XP_005268782.1
XM_011538031.1 Intron XP_011536333.1
XM_011538033.1 Intron XP_011536335.1
XM_011538035.1 Intron XP_011536337.1
XM_011538036.1 Intron XP_011536338.1
XM_011538037.1 Intron XP_011536339.1
XM_011538038.1 Intron XP_011536340.1
XM_011538039.1 Intron XP_011536341.1
XM_011538040.1 Intron XP_011536342.1
XM_011538042.2 Intron XP_011536344.1
XM_017018998.1 Intron XP_016874487.1
XM_017018999.1 Intron XP_016874488.1
XM_017019000.1 Intron XP_016874489.1
XM_017019001.1 Intron XP_016874490.1
XM_017019002.1 Intron XP_016874491.1
XM_017019003.1 Intron XP_016874492.1
XM_017019004.1 Intron XP_016874493.1
XM_017019005.1 Intron XP_016874494.1
XM_017019006.1 Intron XP_016874495.1
XM_017019007.1 Intron XP_016874496.1
XM_017019008.1 Intron XP_016874497.1
XM_017019009.1 Intron XP_016874498.1
XM_017019010.1 Intron XP_016874499.1
XM_017019011.1 Intron XP_016874500.1
XM_017019012.1 Intron XP_016874501.1
XM_017019013.1 Intron XP_016874502.1
XM_017019014.1 Intron XP_016874503.1
XM_017019015.1 Intron XP_016874504.1
XM_017019016.1 Intron XP_016874505.1
XM_017019017.1 Intron XP_016874506.1
XM_017019018.1 Intron XP_016874507.1
XM_017019019.1 Intron XP_016874508.1
XM_017019020.1 Intron XP_016874509.1
XM_017019021.1 Intron XP_016874510.1
XM_017019022.1 Intron XP_016874511.1
XM_017019023.1 Intron XP_016874512.1
XM_017019024.1 Intron XP_016874513.1
XM_017019025.1 Intron XP_016874514.1
XM_017019026.1 Intron XP_016874515.1
XM_017019027.1 Intron XP_016874516.1
XM_017019028.1 Intron XP_016874517.1
XM_017019029.1 Intron XP_016874518.1
XM_017019030.1 Intron XP_016874519.1
XM_017019031.1 Intron XP_016874520.1
XM_017019032.1 Intron XP_016874521.1
XM_017019033.1 Intron XP_016874522.1
Gene
STAC3
Gene Name
SH3 and cysteine rich domain 3
There are no transcripts associated with this gene.

View Full Product Details