Product Details

SNP ID
rs12099847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76345626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCAAATCTTGGCTTCCCCTCTA[A/G]ATTTGTTCCATAAAGTAATTTAATA
Phenotype
MIM: 610148 MIM: 606736
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BBS10 PubMed Links

Gene Details

Gene
BBS10
Gene Name
Bardet-Biedl syndrome 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024685.3 2443 UTR 3 NP_078961.3
Gene
OSBPL8
Gene Name
oxysterol binding protein like 8
There are no transcripts associated with this gene.

View Full Product Details