Product Details

SNP ID
rs11829037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120713290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTCAGATTATTTATGTAAACC[C/T]GAAGACAACATCTACAGTATTGATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4700 PubMed Links

Gene Details

Gene
MIR4700
Gene Name
microRNA 4700
There are no transcripts associated with this gene.

Gene
UNC119B
Gene Name
unc-119 lipid binding chaperone B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080533.2 301 Silent Mutation NP_001074002.1

View Full Product Details