Product Details

SNP ID
rs12327034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:26228488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACCGAATACATTATTTGGCA[C/G]TTCTAAATTGTTATTCTGAGTTGTT
Phenotype
MIM: 601689
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TAF4B PubMed Links

Gene Details

Gene
TAF4B
Gene Name
TATA-box binding protein associated factor 4b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293725.1 Intron NP_001280654.1
NM_005640.2 Intron NP_005631.1
XM_005258339.3 Intron XP_005258396.1
XM_011526153.2 Intron XP_011524455.1
XM_017025932.1 Intron XP_016881421.1
XM_017025933.1 Intron XP_016881422.1

View Full Product Details