Product Details

SNP ID
hCV31591085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100603146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCCTGCCCTTGCCACTCTCCAG[C/G]CCCTTTCCTGAGCTCCCCCCTCCCC
Phenotype
MIM: 615052 MIM: 610350
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASB7 PubMed Links

Gene Details

Gene
ASB7
Gene Name
ankyrin repeat and SOCS box containing 7
There are no transcripts associated with this gene.

Gene
LINS1
Gene Name
lines homolog 1
There are no transcripts associated with this gene.

View Full Product Details