Product Details

SNP ID
rs11752227
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31717612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTTGATTCACCAACATCCAGC[A/C]TGCGTCGCAGCCCATCATTGCAATC
Phenotype
MIM: 606520 MIM: 604744 MIM: 610435 MIM: 606038 MIM: 610437 MIM: 611404
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
C6orf25 PubMed Links
Additional Information
For this assay, SNP(s) [rs118062293] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C6orf25
Gene Name
chromosome 6 open reading frame 25
There are no transcripts associated with this gene.

Gene
DDAH2
Gene Name
dimethylarginine dimethylaminohydrolase 2
There are no transcripts associated with this gene.

Gene
LY6G6C
Gene Name
lymphocyte antigen 6 complex, locus G6C
There are no transcripts associated with this gene.

Gene
LY6G6D
Gene Name
lymphocyte antigen 6 complex, locus G6D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021246.2 210 Silent Mutation GCA,GCC A70A NP_067069.2
Gene
LY6G6E
Gene Name
lymphocyte antigen 6 complex, locus G6E (pseudogene)
There are no transcripts associated with this gene.

Gene
LY6G6F
Gene Name
lymphocyte antigen 6 complex, locus G6F
There are no transcripts associated with this gene.

View Full Product Details