Product Details
- SNP ID
-
hCV31594935
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:174301105 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAACAAAATGCAGTGCTATCATCA[G/T]GTATTTGTTCTCTGTCTTACTAAAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CEP44
PubMed Links
Gene Details
- Gene
- CEP44
- Gene Name
- centrosomal protein 44
There are no transcripts associated with this gene.
View Full Product Details