Product Details

SNP ID
rs4699798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:100399680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGACAAGGCCTCTCCTTCAATTA[C/T]TGAACTAGTTTTTTAGGCATATTTT
Phenotype
MIM: 608350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMCN PubMed Links

Gene Details

Gene
EMCN
Gene Name
endomucin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159694.1 Intron NP_001153166.1
NM_016242.3 Intron NP_057326.2
XM_011532024.2 Intron XP_011530326.1
XM_017008290.1 Intron XP_016863779.1
Gene
SNORA101A
Gene Name
small nucleolar RNA, H/ACA box 101A
There are no transcripts associated with this gene.

View Full Product Details