Product Details

SNP ID
rs12072178
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:53926629 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAATTAAGTCTCTTTTAACCTT[C/G]AACTGCTTTCTCTCCCCTACACACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSPB11 PubMed Links

Gene Details

Gene
HSPB11
Gene Name
heat shock protein family B (small) member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316935.1 Intron NP_001303864.1
NM_016126.3 Intron NP_057210.2

View Full Product Details