Product Details

SNP ID
rs11807478
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:46395645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGGAAGGCTTTACAGGTTGGGA[A/C]CCGCAGGACTCAGGAGGGTGTGCCT
Phenotype
MIM: 602935
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAAH PubMed Links

Gene Details

Gene
FAAH
Gene Name
fatty acid amide hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001441.2 Intron NP_001432.2

View Full Product Details