Product Details

SNP ID
rs11618397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:47947418 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAAAATTGGTCAATAAAGGCAAA[C/T]AATCAGGTAATTATCCTACTTTTCC
Phenotype
MIM: 603921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUCLA2 PubMed Links

Gene Details

Gene
SUCLA2
Gene Name
succinate-CoA ligase ADP-forming beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003850.2 Intron NP_003841.1
XM_017020806.1 Intron XP_016876295.1
XM_017020807.1 Intron XP_016876296.1
XM_017020808.1 Intron XP_016876297.1
XM_017020809.1 Intron XP_016876298.1

View Full Product Details