Product Details

SNP ID
rs11616323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49313708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGACCACATGACCAGAATGCAGA[C/T]CAAAGTGTCTCTGAGCCCTACAGAG
Phenotype
MIM: 612175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAB39L PubMed Links

Gene Details

Gene
CAB39L
Gene Name
calcium binding protein 39 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079670.2 Intron NP_001073138.1
NM_001287337.1 Intron NP_001274266.1
NM_001287338.1 Intron NP_001274267.1
NM_001287339.1 Intron NP_001274268.1
NM_030925.3 Intron NP_112187.2
XM_011535254.2 Intron XP_011533556.1
XM_011535255.2 Intron XP_011533557.1
XM_011535256.2 Intron XP_011533558.1
XM_017020785.1 Intron XP_016876274.1
XM_017020786.1 Intron XP_016876275.1
XM_017020787.1 Intron XP_016876276.1
XM_017020788.1 Intron XP_016876277.1

View Full Product Details