Product Details

SNP ID
rs12189657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:168057364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCGGGGAGAGGCCATGGGATGA[C/G]GCCTGCTGCCCCCTCTCACTCCCAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FRMD1 PubMed Links

Gene Details

Gene
FRMD1
Gene Name
FERM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122841.2 Intron NP_001116313.1
NM_024919.4 Intron NP_079195.3
XM_011536137.1 Intron XP_011534439.1
XM_011536138.1 Intron XP_011534440.1
XM_011536140.1 Intron XP_011534442.1
XM_011536141.1 Intron XP_011534443.1
XM_011536142.2 Intron XP_011534444.1
XM_011536143.1 Intron XP_011534445.1
XM_011536145.1 Intron XP_011534447.1
XM_011536146.2 Intron XP_011534448.1
XM_017011317.1 Intron XP_016866806.1
XM_017011318.1 Intron XP_016866807.1

View Full Product Details