Product Details

SNP ID
rs11701602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38375365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATGCATGTCTTGTCTTGCATTA[G/T]AGATTTTTATGTACATTTTCCCTTT
Phenotype
MIM: 165080
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERG PubMed Links

Gene Details

Gene
ERG
Gene Name
ERG, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136154.1 Intron NP_001129626.1
NM_001136155.1 Intron NP_001129627.1
NM_001243428.1 Intron NP_001230357.1
NM_001243429.1 Intron NP_001230358.1
NM_001243432.2 Intron NP_001230361.1
NM_001291391.1 Intron NP_001278320.1
NM_004449.4 Intron NP_004440.1
NM_182918.3 Intron NP_891548.1
XM_011529486.2 Intron XP_011527788.1
XM_011529487.2 Intron XP_011527789.1
XM_017028287.1 Intron XP_016883776.1
XM_017028288.1 Intron XP_016883777.1
XM_017028289.1 Intron XP_016883778.1

View Full Product Details