Product Details

SNP ID
rs6430585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:135749357 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAAGGACATTTATAGGTTCCTCA[A/C]AGTACCTGATGTCCACAAGTCTATT
Phenotype
MIM: 611216
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107985946 PubMed Links

Gene Details

Gene
LOC107985946
Gene Name
uncharacterized LOC107985946
There are no transcripts associated with this gene.

Gene
UBXN4
Gene Name
UBX domain protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014607.3 Intron NP_055422.1

View Full Product Details