Product Details

SNP ID
rs6745996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:135534855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCTTTTTTGAACTGGTACTTTA[A/G]TATACCTAATTACATCCATTTGAGT
Phenotype
MIM: 615655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
R3HDM1 PubMed Links

Gene Details

Gene
R3HDM1
Gene Name
R3H domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282798.1 Intron NP_001269727.1
NM_001282799.1 Intron NP_001269728.1
NM_001282800.1 Intron NP_001269729.1
NM_015361.3 Intron NP_056176.2
XM_005263626.3 Intron XP_005263683.1
XM_005263627.3 Intron XP_005263684.1
XM_005263628.3 Intron XP_005263685.1
XM_005263629.3 Intron XP_005263686.1
XM_006712389.2 Intron XP_006712452.1
XM_011510870.1 Intron XP_011509172.1
XM_011510871.1 Intron XP_011509173.1
XM_011510872.2 Intron XP_011509174.1
XM_011510873.2 Intron XP_011509175.1
XM_011510874.2 Intron XP_011509176.1
XM_011510875.2 Intron XP_011509177.1
XM_011510876.2 Intron XP_011509178.1
XM_011510877.2 Intron XP_011509179.1
XM_011510878.2 Intron XP_011509180.1
XM_011510879.2 Intron XP_011509181.1
XM_011510881.1 Intron XP_011509183.1
XM_017003718.1 Intron XP_016859207.1
XM_017003719.1 Intron XP_016859208.1
XM_017003720.1 Intron XP_016859209.1
XM_017003721.1 Intron XP_016859210.1
XM_017003722.1 Intron XP_016859211.1
XM_017003723.1 Intron XP_016859212.1
XM_017003724.1 Intron XP_016859213.1
XM_017003725.1 Intron XP_016859214.1
XM_017003726.1 Intron XP_016859215.1
XM_017003727.1 Intron XP_016859216.1
XM_017003728.1 Intron XP_016859217.1
XM_017003729.1 Intron XP_016859218.1
XM_017003730.1 Intron XP_016859219.1
XM_017003731.1 Intron XP_016859220.1
XM_017003732.1 Intron XP_016859221.1
XM_017003733.1 Intron XP_016859222.1
XM_017003734.1 Intron XP_016859223.1
XM_017003735.1 Intron XP_016859224.1
XM_017003736.1 Intron XP_016859225.1
Gene
ZRANB3
Gene Name
zinc finger RANBP2-type containing 3
There are no transcripts associated with this gene.

View Full Product Details