Product Details

SNP ID
rs12387310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:129464814 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGACAACATGACATAATGTGCCT[A/T]CTGATGGGATGCAATAAAAAATACA
Phenotype
MIM: 300012
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SMARCA1 PubMed Links

Gene Details

Gene
SMARCA1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282874.1 Intron NP_001269803.1
NM_001282875.1 Intron NP_001269804.1
NM_003069.4 Intron NP_003060.2
XM_005262461.2 Intron XP_005262518.1
XM_005262462.2 Intron XP_005262519.1
XM_006724782.2 Intron XP_006724845.1
XM_017029750.1 Intron XP_016885239.1
XM_017029751.1 Intron XP_016885240.1

View Full Product Details