Product Details
- SNP ID
-
rs12403865
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:16367508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCCATTTCTCTCCTTTCCTTTGC[A/C]TGGTGCGCCTCCCTCCGCCCACTAA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SZRD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149335138] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SZRD1
- Gene Name
- SUZ RNA binding domain containing 1
View Full Product Details