Product Details

SNP ID
rs13398519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108245432 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTATGCTAAAGCATTTACCCATAG[A/G]AAAAGGGTTCTGGTTAACTTCCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SULT1C3 PubMed Links

Gene Details

Gene
SULT1C3
Gene Name
sulfotransferase family 1C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008743.2 Intron NP_001008743.1
NM_001320878.1 Intron NP_001307807.1
XM_017004153.1 Intron XP_016859642.1
XM_017004154.1 Intron XP_016859643.1
XM_017004155.1 Intron XP_016859644.1

View Full Product Details