Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001198818.1 | 2474 | UTR 3 | NP_001185747.1 | ||
NM_001198819.1 | 2474 | UTR 3 | NP_001185748.1 | ||
NM_001198820.1 | 2474 | UTR 3 | NP_001185749.1 | ||
NM_014576.3 | 2474 | UTR 3 | NP_055391.2 | ||
NM_138932.2 | 2474 | UTR 3 | NP_620310.1 | ||
NM_138933.2 | 2474 | UTR 3 | NP_620311.1 | ||
XM_005269718.2 | 2474 | UTR 3 | XP_005269775.1 | ||
XM_005269720.3 | 2474 | UTR 3 | XP_005269777.1 | ||
XM_011539729.2 | 2474 | UTR 3 | XP_011538031.1 | ||
XM_011539730.2 | 2474 | UTR 3 | XP_011538032.1 | ||
XM_017016160.1 | 2474 | UTR 3 | XP_016871649.1 |