Product Details

SNP ID
rs11864294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:24924245 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAAAGGTTATTTTAGAAAGACAC[C/T]TGTAAGAACCTGGTTTTCTTTAAAC
Phenotype
MIM: 608293
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP17 PubMed Links

Gene Details

Gene
ARHGAP17
Gene Name
Rho GTPase activating protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006634.2 Intron NP_001006635.1
NM_018054.5 Intron NP_060524.4
XM_011545873.2 Intron XP_011544175.1
XM_011545874.2 Intron XP_011544176.1
XM_011545875.2 Intron XP_011544177.1
XM_011545876.2 Intron XP_011544178.1
XM_011545877.2 Intron XP_011544179.1
XM_011545878.2 Intron XP_011544180.1
XM_017023390.1 Intron XP_016878879.1

View Full Product Details