Product Details

SNP ID
rs10941328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37391416 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATACATATTAAACAATTCTTCATT[C/T]CCCCTCCCTCTAGCCTCTGATAACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR70 PubMed Links

Gene Details

Gene
WDR70
Gene Name
WD repeat domain 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018034.2 Intron NP_060504.1
XM_005248318.3 Intron XP_005248375.1
XM_011514061.2 Intron XP_011512363.1
XM_017009604.1 Intron XP_016865093.1
XM_017009605.1 Intron XP_016865094.1

View Full Product Details