Product Details

SNP ID
rs11567700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:35859495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCATACCCTGCTGAAACTACA[C/T]AAAGAGCTTTTGCTGGTGGCAGAAC
Phenotype
MIM: 146661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL7R PubMed Links

Gene Details

Gene
IL7R
Gene Name
interleukin 7 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002185.3 Intron NP_002176.2
XM_005248299.3 Intron XP_005248356.1

View Full Product Details