Product Details

SNP ID
rs12045013
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:1547458 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTACACAAGGATGAGCGAGCAGCC[G/A]CCCTTCTGGATCTCCTGGCGAGGAG
Phenotype
MIM: 616101
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SSU72 PubMed Links

Gene Details

Gene
SSU72
Gene Name
SSU72 homolog, RNA polymerase II CTD phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014188.2 Intron NP_054907.1
Gene
TMEM240
Gene Name
transmembrane protein 240
There are no transcripts associated with this gene.

View Full Product Details