Product Details

SNP ID
rs12730740
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:111548374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTTTGATTGTGCCTTGTCTTTA[G/T]GATCATACTGGTAAAACTATATGTC
Phenotype
MIM: 179520
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAP1A PubMed Links

Gene Details

Gene
RAP1A
Gene Name
RAP1A, member of RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010935.2 Intron NP_001010935.1
NM_001291896.1 Intron NP_001278825.1
NM_002884.3 Intron NP_002875.1
XM_017001960.1 Intron XP_016857449.1
XM_017001961.1 Intron XP_016857450.1
XM_017001962.1 Intron XP_016857451.1
XM_017001963.1 Intron XP_016857452.1
XM_017001964.1 Intron XP_016857453.1
Gene
TMIGD3
Gene Name
transmembrane and immunoglobulin domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081976.2 Intron NP_001075445.1
NM_001302680.1 Intron NP_001289609.1
NM_020683.6 Intron NP_065734.5

View Full Product Details