Product Details

SNP ID
rs12140599
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:111329498 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGATGCCAATGATTCATAAATTC[A/G]TCCTCTTCACATAAACCCATATATA
Phenotype
MIM: 606080 MIM: 614234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHIA PubMed Links

Gene Details

Gene
CHIA
Gene Name
chitinase, acidic
There are no transcripts associated with this gene.

Gene
PIFO
Gene Name
primary cilia formation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300831.1 Intron NP_001287760.1
NM_181643.5 Intron NP_857594.2
XM_005270472.1 Intron XP_005270529.1
XM_017000322.1 Intron XP_016855811.1

View Full Product Details