Product Details
- SNP ID
-
rs13306709
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:41332382 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTGTGTCACCCTAGGTTGCCCCC[C/T]CAGCCCTATCTCCATCTGGGTCTCC
- Phenotype
-
MIM: 190180
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC97
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147219478] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC97
- Gene Name
- coiled-coil domain containing 97
There are no transcripts associated with this gene.
- Gene
- TGFB1
- Gene Name
- transforming growth factor beta 1
View Full Product Details