Product Details

SNP ID
rs12818603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52171236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGGTGATGCTCCTCTCCCACC[C/G]TGGCAGCCCACAAAACACAGTCAGT
Phenotype
MIM: 611161
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRT80 PubMed Links

Gene Details

Gene
KRT80
Gene Name
keratin 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081492.1 1654 UTR 3 NP_001074961.1
NM_182507.2 1654 UTR 3 NP_872313.2
XM_005268676.3 1654 UTR 3 XP_005268733.1

View Full Product Details