Product Details
- SNP ID
-
rs10739281
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:109647530 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTAGATATTGCCCAGGTGCCTCCG[A/G]AAGTATTGTATCCCTTTACATTGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PALM2
PubMed Links
Gene Details
- Gene
- PALM2
- Gene Name
- paralemmin 2
View Full Product Details