Product Details

SNP ID
rs3751849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72783295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCCAAACTTGCCCCCCTCCCC[C/G]CTGAAGAAAGTAAACAAACAAAAAA
Phenotype
MIM: 104155
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZFHX3 PubMed Links

Gene Details

Gene
ZFHX3
Gene Name
zinc finger homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164766.1 12368 UTR 3 NP_001158238.1
NM_006885.3 12368 UTR 3 NP_008816.3
XM_005255957.3 12368 UTR 3 XP_005256014.1
XM_017023251.1 12368 UTR 3 XP_016878740.1

View Full Product Details