Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000790.3 | 1756 | UTR 3 | NP_000781.1 | ||
NM_001082971.1 | 1756 | UTR 3 | NP_001076440.1 | ||
NM_001242886.1 | 1756 | UTR 3 | NP_001229815.1 | ||
NM_001242887.1 | 1756 | UTR 3 | NP_001229816.1 | ||
NM_001242888.1 | 1756 | UTR 3 | NP_001229817.1 | ||
NM_001242889.1 | 1756 | UTR 3 | NP_001229818.1 | ||
NM_001242890.1 | 1756 | Intron | NP_001229819.1 | ||
XM_005271745.4 | 1756 | UTR 3 | XP_005271802.1 | ||
XM_011515161.2 | 1756 | UTR 3 | XP_011513463.2 |