Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001193434.1 | Intron | NP_001180363.1 | |||
NM_001193435.1 | Intron | NP_001180364.1 | |||
NM_024889.4 | Intron | NP_079165.3 | |||
NM_182601.1 | Intron | NP_872407.1 | |||
XM_005270162.2 | Intron | XP_005270219.1 | |||
XM_005270163.2 | Intron | XP_005270220.1 | |||
XM_011540171.1 | Intron | XP_011538473.1 | |||
XM_011540172.1 | Intron | XP_011538474.1 | |||
XM_011540173.1 | Intron | XP_011538475.1 | |||
XM_011540174.2 | Intron | XP_011538476.1 | |||
XM_011540175.1 | Intron | XP_011538477.1 | |||
XM_011540176.1 | Intron | XP_011538478.1 | |||
XM_017016666.1 | Intron | XP_016872155.1 |