Product Details

SNP ID
rs11190082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99397479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAATCCATGGTATGTACATGTA[C/G]GTTTGTGCAGGCAGGGAACACACAT
Phenotype
MIM: 607802 MIM: 138180
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNNM1 PubMed Links

Gene Details

Gene
CNNM1
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 1
There are no transcripts associated with this gene.

Gene
GOT1
Gene Name
glutamic-oxaloacetic transaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002079.2 1518 UTR 3 NP_002070.1

View Full Product Details