Product Details

SNP ID
rs12220731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96307989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAGGCTGGTTTTGGACTCCTA[A/G]CTCCTGCCTTCACCTCCCAAAGGAT
Phenotype
MIM: 187410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNTT PubMed Links

Gene Details

Gene
DNTT
Gene Name
DNA nucleotidylexotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017520.1 Intron NP_001017520.1
NM_004088.3 Intron NP_004079.3

View Full Product Details