Product Details

SNP ID
hCV31658636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95975563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAAAAGTGATGAGCTTACAAAA[C/G]ATTGAAATCAGTGGGGAAAAAGACC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CC2D2B PubMed Links

Gene Details

Gene
CC2D2B
Gene Name
coiled-coil and C2 domain containing 2B
There are no transcripts associated with this gene.

Gene
ENTPD1-AS1
Gene Name
ENTPD1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details