Product Details

SNP ID
rs11188443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95715102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGATTCTTGTTATGCCTTTTT[G/T]ATTTCTCCTGGAACTTCATGATAAC
Phenotype
MIM: 601752
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ENTPD1 PubMed Links

Gene Details

Gene
ENTPD1
Gene Name
ectonucleoside triphosphate diphosphohydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098175.1 Intron NP_001091645.1
NM_001164178.1 Intron NP_001157650.1
NM_001164179.1 Intron NP_001157651.1
NM_001164181.1 Intron NP_001157653.1
NM_001164182.1 Intron NP_001157654.1
NM_001164183.1 Intron NP_001157655.1
NM_001312654.1 Intron NP_001299583.1
NM_001320916.1 Intron NP_001307845.1
NM_001776.5 Intron NP_001767.3
XM_011540370.2 Intron XP_011538672.2
XM_011540371.2 Intron XP_011538673.1
XM_011540372.2 Intron XP_011538674.2
XM_011540373.2 Intron XP_011538675.2
XM_011540374.2 Intron XP_011538676.2
XM_011540376.2 Intron XP_011538678.2
XM_011540377.2 Intron XP_011538679.1
XM_017016958.1 Intron XP_016872447.1
XM_017016959.1 Intron XP_016872448.1
XM_017016960.1 Intron XP_016872449.1
XM_017016961.1 Intron XP_016872450.1
XM_017016962.1 Intron XP_016872451.1
XM_017016963.1 Intron XP_016872452.1
XM_017016964.1 Intron XP_016872453.1

View Full Product Details