Product Details

SNP ID
rs7079243
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89695853 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAAAGGTCTTACATAATACCTTT[C/T]CAGGTCTGATAAGAGACATTTGCCA
Phenotype
MIM: 605498
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FLJ37201 PubMed Links

Gene Details

Gene
FLJ37201
Gene Name
tigger transposable element derived 2 pseudogene
There are no transcripts associated with this gene.

Gene
KIF20B
Gene Name
kinesin family member 20B
There are no transcripts associated with this gene.

View Full Product Details