Product Details

SNP ID
rs11101787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133075855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTGAAAACGGAGGGCAGAGATTT[A/G]GGGGAGGAATTTTAAAGTCAACTTG
Phenotype
MIM: 612302
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRA1 PubMed Links

Gene Details

Gene
ADGRA1
Gene Name
adhesion G protein-coupled receptor A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083909.2 Intron NP_001077378.1
NM_001291085.1 Intron NP_001278014.1
XM_005252695.1 Intron XP_005252752.1
XM_011540273.1 Intron XP_011538575.1
XM_017016779.1 Intron XP_016872268.1
Gene
ADGRA1-AS1
Gene Name
adhesion G protein-coupled receptor A1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details