Product Details

SNP ID
rs12109653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138178994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGAACGACTGGAATTCCAGTCA[A/G]GCCCGCCCCCTTTCCTTACGCGGAT
Phenotype
MIM: 602848 MIM: 603462 MIM: 605664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD8 PubMed Links

Gene Details

Gene
BRD8
Gene Name
bromodomain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164326.1 267 Intron NP_001157798.1
NM_001300961.1 267 Intron NP_001287890.1
NM_001300962.1 267 Intron NP_001287891.1
NM_001300966.1 267 Intron NP_001287895.1
NM_006696.3 267 Intron NP_006687.3
NM_139199.1 267 Intron NP_631938.1
XM_005271855.2 267 Intron XP_005271912.1
XM_005271856.2 267 Intron XP_005271913.1
XM_005271857.2 267 Intron XP_005271914.1
XM_005271859.3 267 Intron XP_005271916.1
XM_005271860.3 267 Intron XP_005271917.1
XM_005271861.3 267 Intron XP_005271918.1
XM_011543109.1 267 Intron XP_011541411.1
XM_011543110.1 267 Intron XP_011541412.1
XM_017008969.1 267 Intron XP_016864458.1
XM_017008970.1 267 Intron XP_016864459.1
XM_017008971.1 267 Intron XP_016864460.1
XM_017008972.1 267 Intron XP_016864461.1
XM_017008973.1 267 Intron XP_016864462.1
XM_017008974.1 267 Intron XP_016864463.1
XM_017008975.1 267 Intron XP_016864464.1
XM_017008976.1 267 Intron XP_016864465.1
Gene
CDC23
Gene Name
cell division cycle 23
There are no transcripts associated with this gene.

Gene
KIF20A
Gene Name
kinesin family member 20A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005733.2 267 UTR 5 NP_005724.1

View Full Product Details