Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_031271.3 | Intron | NP_112561.2 | |||
XM_005273575.3 | Intron | XP_005273632.1 | |||
XM_005273576.3 | Intron | XP_005273633.1 | |||
XM_006716369.3 | Intron | XP_006716432.1 | |||
XM_011544588.2 | Intron | XP_011542890.1 | |||
XM_011544589.2 | Intron | XP_011542891.1 | |||
XM_011544590.2 | Intron | XP_011542892.1 | |||
XM_011544591.2 | Intron | XP_011542893.1 | |||
XM_011544592.2 | Intron | XP_011542894.1 | |||
XM_011544593.2 | Intron | XP_011542895.1 | |||
XM_011544594.2 | Intron | XP_011542896.1 |