Product Details

SNP ID
rs7359931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34263892 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTAAGAGTGAGGACTGTGCAAG[A/G]CGGCTTGGGTTCAAGGCCTGCCCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0355 PubMed Links

Gene Details

Gene
KIAA0355
Gene Name
KIAA0355
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014686.3 Intron NP_055501.2
XM_005259444.1 Intron XP_005259501.1
XM_017027525.1 Intron XP_016883014.1
XM_017027526.1 Intron XP_016883015.1

View Full Product Details