Product Details

SNP ID
rs6443276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9967221 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGGTCTCGCTCTGTCACCCAAG[A/C]TGGTGTGCAGTGGCACAATCACAGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EMC3 PubMed Links

Gene Details

Gene
EMC3
Gene Name
ER membrane protein complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018447.3 Intron NP_060917.1
XM_005265321.3 Intron XP_005265378.1

View Full Product Details