Product Details

SNP ID
rs11465853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10165297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCGCTCTGAGTCCAGAGAACG[C/G]TGGGTACGGGGGCCCTCCTGTCAGC
Phenotype
MIM: 603304
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRAK2 PubMed Links

Gene Details

Gene
IRAK2
Gene Name
interleukin 1 receptor associated kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001570.3 Intron NP_001561.3

View Full Product Details