Product Details

SNP ID
rs12634007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53287590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATTCTGCTCCAGTCATAGGTTG[G/T]GGTTGTCTTTGTTCTTGGTCAGAAC
Phenotype
MIM: 607010
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DCP1A PubMed Links

Gene Details

Gene
DCP1A
Gene Name
decapping mRNA 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290204.1 1735 Missense Mutation CAC,CCC H542P NP_001277133.1
NM_001290205.1 1735 Missense Mutation CAC,CCC H508P NP_001277134.1
NM_001290206.1 1735 Missense Mutation CAC,CCC H459P NP_001277135.1
NM_001290207.1 1735 Missense Mutation CAC,CCC H421P NP_001277136.1
NM_018403.6 1735 Missense Mutation CAC,CCC H580P NP_060873.4

View Full Product Details