Product Details
- SNP ID
-
rs13332449
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89660411 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGGGGAAGGAGGACCGCCTGTTG[A/G]AAGGGCCCCAGCAGTGTCTGTCACA
- Phenotype
-
MIM: 164010
MIM: 615409
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHMP1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79088174] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHMP1A
- Gene Name
- charged multivesicular body protein 1A
There are no transcripts associated with this gene.
- Gene
- SPATA33
- Gene Name
- spermatogenesis associated 33
View Full Product Details