Product Details

SNP ID
rs3802876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117902112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACACACACACACACAGAGGCAG[C/T]AGACAGTGGAGGTGGGAGTCCGATG
Phenotype
MIM: 610050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS13 PubMed Links

Gene Details

Gene
TMPRSS13
Gene Name
transmembrane protease, serine 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077263.2 1872 UTR 3 NP_001070731.1
NM_001206789.1 1872 UTR 3 NP_001193718.1
NM_001206790.1 1872 Intron NP_001193719.1
NM_001244995.1 1872 Intron NP_001231924.1

View Full Product Details