Product Details

SNP ID
rs12270557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:121457069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCGGGGCCTTGGGTTGGCTCA[A/G]TAAGGGACGTGACCTTGAAAAGCTC
Phenotype
MIM: 602005
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105369536 PubMed Links

Gene Details

Gene
LOC105369536
Gene Name
uncharacterized LOC105369536
There are no transcripts associated with this gene.

Gene
SORL1
Gene Name
sortilin related receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003105.5 Intron NP_003096.1
XM_011542963.2 Intron XP_011541265.1
XM_011542965.2 Intron XP_011541267.1
XM_011542967.2 Intron XP_011541269.1
XM_017018169.1 Intron XP_016873658.1
XM_017018170.1 Intron XP_016873659.1
XM_017018171.1 Intron XP_016873660.1
XM_017018172.1 Intron XP_016873661.1

View Full Product Details