Product Details

SNP ID
rs11029380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26195290 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTAGCAACATGCAACAGTGTTC[C/T]GAAGATATCAGGAGTCCTTGTAAAG
Phenotype
MIM: 610110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANO3 PubMed Links

Gene Details

Gene
ANO3
Gene Name
anoctamin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313726.1 Intron NP_001300655.1
NM_001313727.1 Intron NP_001300656.1
NM_031418.3 Intron NP_113606.2
XM_011520282.2 Intron XP_011518584.1
XM_017018118.1 Intron XP_016873607.1
XM_017018119.1 Intron XP_016873608.1

View Full Product Details