Product Details

SNP ID
rs11023423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:15074184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGCATCTGCTGCTAAAACGCG[C/T]TCCGGCAGAGGGCTGGCTTTGCGGG
Phenotype
MIM: 114160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALCB PubMed Links

Gene Details

Gene
CALCB
Gene Name
calcitonin related polypeptide beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000728.3 Intron NP_000719.1

View Full Product Details